United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 13
of
18 >
EW-33855-45
Muffle Furnace, SSP, 1750 cu. Ft.
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33855-45
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Furnaces
>
Muffle Furnaces
>
Neytech Muffle Furnaces
Share
|
Furnaces with SSP control; 1750 cu in; overall dimensions; 24" x 21 1/2" x 21"; 4600 watts; 200-250V
EW-33855-45
$3,700.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Combined fiber and firebrick insulation cools quickly and provides extended heating element life
Heating elements slide out for quick replacement
Vertical lift door directs heat away from user and saves counter space
All models feature fiber insulation in the chamber and firebrick insulation around the heating elements. The fiber insulation cools down fast while the firebrick provides longer life for the heating elements. Heating elements are easily replaceable—just disconnect and slide out.
Vertical lift door directs heat away from the operator and has maximum access with minimum head room for easy loading and unloading. Protect the bottom heating element with a ceramic hearth plate
Single Set Point Control Furnaces
have proportional
temperature control and include an analog temperature indicator with 50°C/°F increments.
What's Included:
a 5-ft (1.5-m) power cord with a three-prong grounded plug.
Specifications
Product Type
Muffle Furnaces
Minimum temperature (º C)
200
Maximum temperature (º C)
1100
Minimum temperature (º F)
392
Maximum temperature (º F)
2012
Temp control accuracy
±0.25°C
Rise time
45 min
Temp uniformity
±1.9°C at 100°C
Chamber size (" H)
10
Chamber size (" D)
12.5
Manufacturer number
9493407
Power (VAC)
200-250
Power (Hz)
50/60
Model
9493407
Brand
Neytech
Accessories
Qty
/
Price
Availability
Ceramic Hearth Plate for 1750 cu in Furnace with Single Set Point Control
EW-33855-72
at
$171.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title