dummy
EW-23000-38
Call 1-888-358-4705 for availability.
Qty:
- A Compact and Powerful Electrical Calibrator
- Measure, source and simulate multiple parameters
- HART digital configurator
- Easy-to-use, with video quality touch screen, which can be operated by a gloved hand
- Weather-proof to IP 65
- Can display up to 6 active readings
- Available with a range of add-on modules, pressure measurement modules and generation stations, Intecal Field Calibration documenting module, 300Vac measurement probe, wireless (IEEE 802.11g) adaptor
- Digital interface for pressure modules and future options
- USB master and slave for connecting to a PC and peripherals
- Available in Windows CE version
- Long-life Lithium-Polymer battery pack
The DPI 620 electrical calibrator can measure and source mA, mV, V, Ohms, frequency and a variety of RTDs and T/Cs. It provides an isolated 24V loop power supply to energise devices and control loops and a stabilized dc voltage supply for ratio metric transducers. A HART digital configurator is provided for set-up and calibration adjustment of HART devices.
The high resolution, colour touch screen of the calibrator shows only those buttons necessary for the immediate, selected tasks. The buttons have been sized so that they can be operated by a gloved hand, with no need for a stylus. The display can be configured to show up to six reading windows and each window can show an electrical input/output, pressure, temperature, a USB-connected device or a wireless-connected device.
What's Included:
Includes an integral HART communicator, USB interface, rechargeable lithium polymer battery, universal mains adaptor/charger, test leads and calibration certificate.
| Product Type |
Multi-function Calibrators |
| Battery life |
Lithium-Polymer battery (included) - 12 hours continuous use |
| Dimensions |
4.5 in Wx 1.7 in H x 7.2 in L (114 x 42 x 183mm) |
| CE Compliance |
Yes |
| Brand |
Druck |
| Manufacturer number |
DPI 620 CE |
| Model |
DPI 620 CE |
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,357.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,357.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,357.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,357.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,593.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,776.00 USD/EACH
Call 1-888-358-4705 for availability.
at $2,139.00 USD/EACH
Call 1-888-358-4705 for availability.
at $442.00 USD/EACH
Call 1-888-358-4705 for availability.
at $803.00 USD/EACH
Call 1-888-358-4705 for availability.
at $59.00 USD/EACH
Call 1-888-358-4705 for availability.
at $96.00 USD/EACH
Call 1-888-358-4705 for availability.
at $78.00 USD/EACH
Call 1-888-358-4705 for availability.
at $774.00 USD/EACH
Call 1-888-358-4705 for availability.
at $204.00 USD/EACH
Usually ships in 14 days.
at $84.00 USD/EACH
Call 1-888-358-4705 for availability.
at $65.00 USD/EACH
Call 1-888-358-4705 for availability.
at $104.00 USD/EACH
Call 1-888-358-4705 for availability.
at $425.00 USD/EACH
Usually ships in 10 days.
Technical Resources
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.