United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
8 >
Representative photo only
GF Signet Conductivity Cell with Integral Electronics K 1 0 4 to 20 mA output 3 4 NPT threads
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-19007-06
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-30
NIST-Traceable Calibration; Conductivity/Resistivity Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Electrodes, Industrial
>
Conductivity Electrodes - Industrial
>
+GF+ Signet Conductivity/Resistivity Electrodes with Integral Electronics
Share
|
+GF+ Signet Conductivity Cell with Integral Electronics, K = 1.0, 4 to 20 mA output, 3/4" NPT threads
EW-19007-06
$804.00 USD/EACH
Usually ships in 10 days.
Qty
:
Add to Cart
Save to shopping list
Integral mount systems for quick and easy installation
Compact design for maximum installation flexibility
Digital (S3L) interface or two-wire 4 to 20 mA output
EasyCal with automatic test solution recognition
+GF+ Signet Conductivity/Resistivity Sensors with Integral Electronics are available in various configurations for maximum installation flexibility. These combined integral system configurations allow for in-line mounting and include conductivity electrodes in a choice of K = 0.01, 0.1, 1.0, or 10.0 cell constants. Users have a choice of two outputs: digital (S3L) or 4 to 20 mA. The digital (S3L) output version allows for up to six sensor inputs directly into the Signet 8900 Multi-Parameter Controller. The two-wire 4 to 20 mA output is available with eight 4 to 20 mA output ranges for each electrode cell constant. Additionally, each range can be inverted and are field selectable by the user.
All units are built with NEMA 4X/IP 65 enclosures which allow wiring connections with long cable runs of up to 1,000 feet (305 m). EasyCal is a standard feature that automatically recognizes conductivity test solution values for simple field calibration.
Specifications
Product Type
Conductivity Cell
Cell constant
K = 1.0
Sensor material
316 Stainless Steel
Temp range
-10 to 85°C (14 to 185°F)
Max temperature (°C)
85
Min temperature (°C)
10
Fitting
3/4" NPT(M)
Brand
+GF+ Signet
Manufacturer number
3-2850-52-41
Model
3-2850-52-41
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Conductivity/Resistivity Meter
EW-17090-30
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual (972 KB)
Related technical articles (Roll over for summary)
Tech Challenge: Faulty pH Readings
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title