log in or register
GF+ Signet Single-Channel Transmitter, Field Mount
EW-56560-14
$531.00 USD/EACH
Usually ships in 13 days.
Qty
  • Add-on modules help you create your own customized transmitter
  • Large, backlit LCD with ''Dial-type" digital bar graph
  • Customizable label and bar graph settings
  • User-friendly interface consistent with legacy instruments
  • Optional plug-in modules easily expand capibilities
These versatile, single-channel transmitters are designed to help you tailor a system to your exact needs. Transmitters feature a large LCD with digital bar graph as well as displays of the main and secondary measurements. Preset values offer quick and easy programming. Or customize your unit for labeling, min/max dial settings, and unit/decimal measurement options. Both panel- and field-mount models run on 12 to 32 VCD power and can be looped with compatible sensors.
For added flexibility, add-on modules let you create the ideal control center. The relay module 56560-15 (for panel mount transmitter only) features two programmable dry-contact relays for hysteresis and time delay. PC COMM module 56560-19 lets you easily configure your transmitter from your computer—save settings and use for future installations. Choose the H COMM module 56560-17 to communicate with any HART®-enabled device and remotely adjust 4 to 20 mA settings.
Expand your measurement capabilities using the optional parameter modules for applications including pH, ORP, temperature, flow, pressure, and level control. Contact our Application Specialists for details.
Product Type Controller
Range (pH) 0.00 - 15.00
Range (Temperature) 99 to 350°C
Range (mV) +/- 1999.9
Accuracy (pH) +/- 0.5% of reading
Output (Current) One 4 to 20 mA, isolated and reversible
Output (Relay) Optional two SPDT relays
Dimensions 3 3/4" x 3 3/4" x 2 1/8"
Control type On/off (limit)
Password protection Yes
Display 7- and 14-segmented LCD, backlit
Power 12 to 32 VDC
Housing 1/4-DIN
CE Compliance Yes
Brand +GF+ Signet
Accessories
Qty/Price
Availability
Universal Mount Kit
EW-05631-50
at $62.50 USD/EACH
In stock.
at $23.00 USD/PACKAGE
Usually ships in 14 days.
Relay Module
EW-56560-15
at $104.00 USD/EACH
Usually ships in 13 days.
at $203.00 USD/EACH
Usually ships in 13 days.
H Comm Module
EW-56560-17
at $276.00 USD/EACH
Usually ships in 13 days.
at $240.00 USD/EACH
Usually ships in 13 days.
at $88.50 USD/EACH
Usually ships in 14 days.
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 9 days.
at $115.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title