log in or register
GammaGuard Sterile Cleanroom Coveralls, double extra large, 25/case
EW-33684-34
$280.00 USD/CASE
Usually ships in 5 days.
Qty
  • Bound seams and tunnelized elastic provide low linting and strong tear resistance
Coveralls feature a sterility assurance of 10-6, low linting, and a more breathable liquid barrier. Special fold design protects against potential contamination during gowning. Feature bound seams and elastic wrists and ankles. Manufactured in an ISO certified facility, clean processed, and individually packaged under cleanroom conditions. Recyclable.
Compliance: ISO Class 4
Product Type Coveralls with elastic wrists and ankles
Description Sterile, recyclable
Size XX-Large
Compliance ISO Class 4
Quantity 25/case
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title