United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
GammaGuard Sterile Cleanroom Coveralls large 25 case
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33684-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Safety Apparel
>
Coveralls
>
GammaGuard Sterile Cleanroom Coveralls
Share
|
GammaGuard Sterile Cleanroom Coveralls, large, 25/case
EW-33684-32
$280.00 USD/CASE
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Bound seams and tunnelized elastic provide low linting and strong tear resistance
Coveralls feature a sterility assurance of 10
-6
, low linting, and a more breathable liquid barrier. Special fold design protects against potential contamination during gowning. Feature bound seams and elastic wrists and ankles. Manufactured in an ISO certified facility, clean processed, and individually packaged under cleanroom conditions. Recyclable.
Compliance:
ISO Class 4
Specifications
Product Type
Coveralls with elastic wrists and ankles
Description
Sterile, recyclable
Size
Large
Compliance
ISO Class 4
Quantity
25/case
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Keeping your Workforce Safe and Healthy Through the Holidays
Chemical Compatibility Chart for Safety Apparel
Safety Apparel Material Selection Guide
Show All...
Cleanroom Garments
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title