log in or register
GasAlertQuattro 4-Gas Detector, Rechargeable, %LEL, O2, H2S, CO
EW-86418-62
$755.00 USD/EACH
Usually ships in 7 days or sooner.
Qty
  • IntelliFlash green LED verifies operation and compliance to user and supervisor
  • Bright, extended visual alarm bars for viewing at any angle
Compact and lightweight design makes these gas detectors comfortable to wear all day. Continuous LCD displays real-time gas concentrations. Built-in concussion-proof boot and IP66/67 rating makes these detectors suitable for field use. Verify operation from a distance by looking for the IntelliFlash green LED with user-defined flash frequency. Additional features include one-button operation, data/event logging, audible/visual/vibrating alarms, user adjustable alarm set points, sensor self-test, and multilanguage capability (English, French, German, Spanish, and Portuguese). Approvals: CE; CSA Class I, Div. 1, Groups A, B, C, D; ATEX
What's Included:
calibration cap and tubing, and either rechargeable Li battery pack or three AA alkaline batteries depending on model. Models with rechargeable battery pack also include a battery charger.
Product Type 4 Gas Monitors/Detectors
Gasses Measured O2, CO, H2S, Combustibles (LEL)
Range (CO) 0 to 1000 ppm
Range (O2) 0 to 30.0%
Range (H2S) 0 to 200 ppm
Range (Combustibles) 0 to 100%
CE Compliance Yes
Alarm six red LEDs visual alarm, vibrating alarm, 95dB audible alarm
Power lithium rechargeable batteries
Brand GasAlertQuattro
Manufacturer number QT-XWHM-R-Y-NA
Accessories
Qty/Price
Availability
at $625.00 USD/EACH
Usually ships in 7 days.
at $195.00 USD/EACH
In stock.
Concussion proof boot
EW-86419-28
at $35.00 USD/EACH
Usually ships in 7 days.
at $350.00 USD/EACH
Usually ships in 7 days.
Manual aspirator pump
EW-86419-32
at $80.00 USD/EACH
Usually ships in 7 days.
at $170.00 USD/EACH
Usually ships in 7 days.
at $150.00 USD/EACH
Usually ships in 7 days.
at $140.00 USD/EACH
Usually ships in 7 days.
Replacement CO sensor
EW-86419-40
at $225.00 USD/EACH
Usually ships in 7 days.
Calibrations
Qty/Price
Availability
at $135.00 USD/EACH
Usually ships in 8 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title