United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 10
of
31 >
Representative photo only
Gauge Guard SS 1 4 NPT f X 1 4 NPT f VAC To 500 PSI
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-68340-18
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Gauge guards, Seals, and Accessories
>
Gauge Guards
Share
|
Gauge Guard, SS, 1/4" NPT (f) X 1/4" NPT (f), VAC To 500 PSI
EW-68340-18
$73.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Protect any gauge with a NPT(M) connector from contaminated, corrosive, hot, or viscous media
Protect transducers, switches, and gauges 3 1/2'' in diameter or smaller
Fill/bleed connection is standard
Gauge Guards are used to isolate and protect pressure or vacuum instruments used on ultra-pure or highly-corrosive fluid lines, whenever such fluids will corrode metal instrument components. Gauge Guards also protect against clogging of instruments by suspended solids or highly viscous fluids. Furthermore, they eliminate dead pocket areas within instruments where chemicals or organic matter could decompose. Gauge guards are a very cost-effective way to protect your very expensive transducers, gauges, or switches.
Process connections are 1/4'' and 1/2'' NPT (F), and 1 1/2'' and 2'' sanitary Tri-Clamp.
Specifications
Product Type
Pressure gauge accessories
Process connection
1/4" NPT(F)
Brand
Cole-Parmer
Manufacturer number
KC05-100
Technical Resources
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title