Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

General Purpose Refrigerator, 17 cu ft

EW-44765-20
$2,487.00 USD/EACH

Usually ships in 12 days.
Qty

  • Durable commercial cooling at an affordable price
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.General Purpose Models are ideal for basic storage of chemicals and reagents. These units are available in undercounter and large capacity models.
What's Included:
Commercial power cord and plug, except for explosion-proof models which require hard-wiring.
Product Type Upright Refrigerators
Capacity (cu ft) 17.5
Capacity (Liters) 496
Dimensions (interior) 56 1/2"H x 26"W x 22 1/2"D
Dimensions (overall) 66 1/2"H x 30"W x 27 1/2"D
Min Temp - Refrigerator (º C) 1
Min Temp - Refrigerator (º F) 33
Max Temp - Refrigerator (º C) 5
Max Temp - Refrigerator (º F) 41
No. of doors 1
No. of shelves 4 adjustable shelves, 1 stationary shelf, 3 storage bins, 5 door shelves
Power (Hz) 60
Brand Marvel
Manufacturer number 17CAR001
Power (VAC) 115





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title