log in or register
Gerber-method Butyrometer; milk test
EW-17425-10
$26.25 USD/EACH
  • Get test results in 15 minutes—saves you time and money
Use these butyrometers to measure the percent fat in dairy products. Butyrometers are precision calibrated and feature permanent graduations. Made of resistant borosilicate glass.
Order glass cups for Roeder and Van-Gulik method testing. Stoppers with special acid resistant rubber are available for all butyrometers. Both stopper 17425-15 and stopper pusher 17425-16 are required for milk and Kohler method testing; stoppers 17425-14 and -17 are required for Roeder and Van-Gulik method testing.
Product Type Extraction bottles
Material Glass
Capacity (Liters) 0.011
Capacity (mL) 11
Accessories
Qty/Price
Availability
at $747.00 USD/BOX of 100
Call 1-888-358-4705 for availability.
at $26.25 USD/BOX of 5
Call 1-888-358-4705 for availability.
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title