log in or register
Gilmont Fallng Ball Viscometer, 0.2 to 2 / 1 to 10 centipoise
EW-08701-00
$238.00 USD/EACH
  • Ideal for routine plant quality control checks and research laboratory applications
This viscometer is extremely easy to use; first fill the tube with sample, then invert tube. Next, restore tube to nornal position, turn the knob to release the ball, and then simply measure the time of descent. The ball moves against a white background with red reference lines making it easy to read. Repeatability with good technique varies from ±0.2 to ±1.0%, depending upon time of descent. Viscometers require a 7-mL sample volume.
Viscometers have wetted materials of glass, Viton®, and Delrin® acetal. PTFE O-rings are available for use with lquids that adhere to Viton®; call our Applications Specialists for more information.
What's Included:
one glass and one 316 stainless steel (SS) high-precision ball. For higher viscosity ranges, use the optional tantalum ball (sold separately).
Hot Tips:
NT falling ball viscometers conform to ASTM D 1343-93.
Product Type Falling Ball Viscometer
Range 0.2 to 2 cp (glass ball)
1 to 10 cp (SS ball)
2 to 20 cp (optional tantalum ball)
Brand Gilmont
Manufacturer number GV-2100
Model GV-2100
Accessories
Qty/Price
Availability
at $213.00 USD/EACH
In stock.
Reconditioned & Like New
Qty/Price
Availability
RECOND 08701-00
EW-00153LX
at $93.00 USD/EACH
In stock.
Technical Resources
  • Instruction Manual
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title