GreenLight® Environmental Series 910 Bateria Counting System
EW-99561-23
$9,995.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 13 days.
Qty
  • Calculate live aerobic bacteria loads in as little as 45 minutes
  • Low cost method of measuring disinfection efficiency and contamination levels
  • Results meet 40 CFR part 160 Good Laboratory Practice Standards
The GreenLight system determines the level of live aerobic bacteria by measuring oxygen consumption. Ideal to monitor wastewater discharge for disinfection limits in NPDES permits; drinking water plant disinfection, or the bacteria loading at public and private beaches.
GreenLight measures dissolved oxygen with a single-use flourescence sensor vial to determine the rate of bacteria respiration. System has a built-in vial incubator and is able to provide results in as little as 45 minutes. Results calculated follwing Standard Methods method 2710 B providing Specific Oxygen Uptake Rate (SOUR) that is correlated to active bacteria as Colony Forming Units (CFU per 100 mL).
Model 910 is a single test system for 15-mL vials.
What's Included:
GreenLight software, and pack of 24 APCheck 15-mL single-use sensor vials.
Product Type Dissolved 0xygen, BOD, and COD
Meter style Benchtop
Dimensions (" W) 3
Dimensions (" H) 8
Dimensions (" D) 10
Connections USB
Power USB
CE Compliance Yes
Brand BASELINE MOCON
Model GREENLINE 910
Accessories
Qty/Price
Availability
at $150.00 USD/PACKAGE
Usually ships in 8 days.
at $192.00 USD/PACKAGE
Usually ships in 8 days.
at $495.00 USD/PACKAGE
Usually ships in 8 days.
at $765.00 USD/PACKAGE
Usually ships in 8 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title