Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
HACH 43975-00 Turbidity Sample Degassing Kit
Hach  |   Mfr# 43975-00  |   Item# EW-99512-68
$55.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 7 days.
Qty
    Kit includes plastic syringe with male slip tip luer and one-holed rubber stoppers to fit 2100 series turbidity meter vials.
    Use is simple. Place sample into vial, put supplied rubber stopper on to the vial, insert syringe (with plunger compressed) into hole of stopper, pull up syringe plunger in order to pull a vacuum on the contents of the vial, hold for 10 seconds, remove syringe, and test sample.
    Product Type Sample processing
    Qty/ea 1
    Brand Hach
    Manufacturer number 43975-00
    Model 43975-00
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      What is included in the kit? Are there pictures of it somewhere?
      Asked on 1/21/2010 by Anonymous
      1 answer
      • STAFF EXPERT
        A:
        The kit includes a syringe, stoppers, and catheter. We do have a picture on file. Please contact us directly (phone or email) for electronic file requests. AnswerBox does not currently have the functionality to send files
        Answered on 1/25/2010 by Jen from Technical Support
    Displaying question 1
    Do you have a question about this product? 
    Product Reviews