United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 24
of
30 >
Representative photo only
H B Instrument Hydrometer 1 200 to 1 420 Specific Gravity 0 002 Divisions Plain Form
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-08298-86
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17107-00
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
$130.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Hydrometers
>
Hydrometers for Specific Gravity
>
H-B Instrument Specific Gravity Plain Form Hydrometers for Liquids Heavier than Water
Share
|
H-B Instrument Hydrometer, 1.200 to 1.420 Specific Gravity, 0.002 Divisions, Plain Form
EW-08298-86
$21.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
NIST Traceable
Temperature of Standardization 60°F/60°F
Metal Ballast
Individually Serialized
Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Specifications
Specific gravity range (Minimum)
1.2
Product Type
Hydrometers, Specific Gravity
Specific gravity range (Maximum)
1.42
Manufacturer number
50350
Form
Plain
Model
50350
Brand
H-B Instrument Company
Accessories
Qty
/
Price
Availability
Hydrometer Case for 190 mm Length Hygrometer
EW-08285-98
at
$39.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
EW-17107-00
at $130.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Hydrometer Selection Guide
How to Read a Hydrometer
Brix Scale (sucrose)
Show All...
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title