United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-99511-10
Hach
®
2100Q Handheld Turbidity Meter, EPA compliant
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-99511-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Turbidity Meters and Turbidity Testing Products
>
Turbidity Handheld Meters
>
Hach 2100Q Handheld Turbidity Meter
Share
|
Hach 2100Q Handheld Turbidity Meter, EPA compliant
EW-99511-10
$1,171.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Easy-to-follow interface with on-screen prompts simplifies calibration and verification
Innovative Rapidly Settling Turbidity
(RST) mode allows for accurate and repeatable measurement of rapidly settling sampleseliminating guesswork or duplicate runs
Customizable power and connectivity options add versatility
Meets or exceeds design and performance criteria as specified in U.S. EPA method 180.1
This portable turbidity meter features a unique optical system that compensates for potential interferences, making it ideal for field or lab use. Intuitive user interface prompts you during calibration and verification for hassle-free setup. On-screen assistance eliminates the need to page through manuals for help.
Unique RST mode reduces measurement uncertainty caused by changing turbidity values in settling samples. RST mode measurements are ultimately more accurate and repeatable than those obtained by traditional smoothing techniques, such as averaging.
Additional meter features include 23 pre-programmed menu languages, time-and-date stamped readings, and closed cap control which helps eliminate stray light interference. Optional power and output modules allow for user customizability.
What's Included:
StablCal™ primary calibration standards in 1" sealed vials (20, 100, 800 NTU), six sample cells, 10 NTU primary verification standard, silicone oil, oiling cloth, four alkaline AA batteries, and carrying case.
Specifications
Product Type
Turbidity Meter
Meter style
Portable
Dimensions (" W)
4-1/4
Dimensions (" H)
3
Dimensions (" L)
9
Range (Turbidity)
0.01 to 1000
Range (Temperature °C)
0 to 50
Range (Temperature °F)
32 to 122
Resolution (Turbidity)
0.01, 0.1, 1
Accuracy (Turbidity)
± 2% of reading
Display
LCD
Interface
USB
Power
Four AA batteries (included)
Datalogging (points)
500
CE Compliance
Yes
Brand
Hach
Model
2100Q Portable Turbidity Meter
Accessories
Qty
/
Price
Availability
USB and Power Module for the 2100Q
EW-99511-13
at
$363.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Power Module for the 2100Q
EW-99511-14
at
$134.00 USD/EACH
In stock.
Add to Cart
Sample Filtration and Degassing Kit
EW-99511-16
at
$80.50 USD/EACH
Usually ships in 4 days.
Add to Cart
StablCal 100 mL calibration kit for the 2100Q
EW-99511-17
at
$168.00 USD/EACH
Usually ships in 4 days.
Add to Cart
StablCal 500 mL calibration kit for the 2100Q
EW-99511-18
at
$287.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Gelex Secondary Standard Set
EW-99511-19
at
$184.10 USD/EACH
Usually ships in 4 days.
Add to Cart
StablCal ampule calibration kit for the 2100Q
EW-99511-20
at
$177.00 USD/EACH
Usually ships in 6 days or sooner.
Add to Cart
10 NTU verification standard
EW-99511-21
at
$20.00 USD/EACH
In stock.
Add to Cart
Replacement sample cells for 99511-00. Pack of six.
EW-99511-55
at
$34.00 USD/PACKAGE
In stock.
Add to Cart
Degassing kit
EW-99512-68
at
$40.75 USD/EACH
Usually ships in 7 days.
Add to Cart
Sample cell oiling cloth
EW-99512-72
at
$14.80 USD/EACH
In stock.
Add to Cart
Replacement silicone oil, 15-mL.
EW-99512-75
at
$17.25 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (920 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Good Laboratory Practices for Taking Turbidity Measurements
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title