United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-99574-60
Hach COD reactor, 115 VAC, 50/60Hz
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-99574-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Dissolved Oxygen, BOD, and COD
>
Chemical Oxygen Demand Testing
>
Chemical Oxygen Demand (COD) Test Systems
>
Hach COD Reactors
Share
|
Hach COD reactor, 115 VAC, 50/60Hz
EW-99574-60
$860.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for wastewater influent and effluent testing, industrial process water testing, and environmental monitoring
Uses EPA-approved COD methodrequires less equipment, lab space, and maintenance than other open reflux COD tests
High flexibility, customization, and temperature accuracy
For a complete system order COD reactor (99574-60 or -65), COD or TOC test kits (99574-25,-26,-27,-28, or -29), and colorimeter (99573-00)
Hach DRB-200 COD Reactor digests up to 15 samples simultaneously. Reactors feature one-touch operation through the use of preprogrammed Hach COD and TNT standard digestion temperatures/times, or customize up to three programs. To customize, choose from one of nine languages, set the temperature between 37 to 165°C, and enter the time between 1‑minute to 8 hours. Built-in timer features audible alarm and automatic shut-off to prevent overheating. Transparent, locking lid covers 16 x 100-mm vials while protecting the user in the unlikely event of a broken reagent vial.
For full specifications and operating instructions view the instruction manual linked below.
Specifications
Product Type
Oxygen Test Kits
Meter style
Test kits
Accessories
Qty
/
Price
Availability
Hach DR/870 Colorimeter
EW-99573-00
at
$1,024.00 USD/EACH
In stock.
Add to Cart
Hach COD Test Kit, Low Range, 0 to 150 ppm
EW-99574-25
at
$38.80 USD/EACH
In stock.
Add to Cart
Hach COD Test Kit, High Range, 0 to 1500 ppm
EW-99574-26
at
$38.80 USD/EACH
In stock.
Add to Cart
Hach COD Test Kit, Manganese III, 20 to 1000 ppm
EW-99574-27
at
$42.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Hach TOC Test Kit, Low Range, 0 to 20 mg/L
EW-99574-28
at
$399.00 USD/EACH
In stock.
Add to Cart
Hach TOC Test Kit, High Range, 20 to 700 mg/L
EW-99574-29
at
$399.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Volumetric pipette, 2.00 mL
EW-99574-52
at
$10.75 USD/EACH
Usually ships in 6 days.
Add to Cart
Pipette filler, safety bulb
EW-99574-53
at
$33.10 USD/EACH
Usually ships in 6 days.
Add to Cart
Test tube rack
EW-99574-54
at
$67.50 USD/EACH
Usually ships in 5 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Dissolved Oxygen (DO) Measurement FAQs
Oxygen/Dissolved Oxygen
Testing the pH of Wastewater Samples
Show All...
Testing the pH of Soil Samples
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title