Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Hach Colorimeter Test Kit, fluoride (SPADNS), 0 to 2.0 ppm, 25/kit
Hach  |   Mfr# 25060-25  |   Item# EW-99574-08
$50.75 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Environmental test kits for your Hach Colorimeter
Order the test kits for the Hach colorimeter to complete your water testing protocol. More test kits are available.
All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120VAC 0r 99574-65, 240 VAC shown below).
This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Water Testing Kit
Tests/kit 25
Qty/ea 1
Brand Hach
Manufacturer number 25060-25
Model 25060-25
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    3 Questions | 3 Answers
    Displaying questions 1-3
    • Q:
      is there 25 testing strips in one package?
      Asked on 2/18/2011 by albert from miami fl
      1 answer
      • STAFF EXPERT
        A:
        You will have everything you need in order to do 25 tests with each of these test kits. All reagents in the test kit are packaged as powder pillows.
        These test kits are meant to be used with the Hach DR/870 Colorimeter (99573-00). This colorimeter does not use test strips.
        Answered on 2/19/2011 by Jaime from Technical Support
    • Q:
      can i use this kit for finding the fluoride content in pastes.I have a colorimeter.
      Asked on 1/13/2011 by lalu from kerala
      1 answer
      • Staff Reviewer
        A:
        This kit has only been used to test water based solutions. If you take a small portion or paste and mix it into water to complete the sample volume this may work for you. However, we do not have an exact method or amount to mix in our suggest.
        Answered on 1/14/2011 by Jen from Technical Support
    • Q:
      I am looking for a home tap water test kit to measure levels of fluoride and heavy metals. Is this the right one?
      Asked on 7/29/2010 by Shoshone
      1 answer
      • STAFF EXPERT
        A:
        This is the kit that contains the test reagents required for colorimetric testing of water for fluoride. To take the measurements one will require a colorimeter such as the 99573-00.
        Answered on 8/2/2010 by Josh from Technical Support
    Displaying questions 1-3
    Do you have a question about this product? 
    Product Reviews