log in or register
Hach DR/870 Colorimeter
EW-99573-00
$1,024.00 USD/EACH
  • Faster to run samples in series—retains the selected method until a different method is selected
  • Performs tests for 70 different parameters
  • Automatically selects the wavelength for each parameter
  • Waterproof housing is IP67 rated and permits on-site testing
This colorimeter combines state-of-the-art features and field readiness with ease of use. The large LCD steps you through set up and procedures and indicates program number, sample number, parameter, test results, unit of measure, and more. Use prepared reagent test kits (sold separately) to run tests quickly—colorimeter comes preprogrammed for 70 parameters routinely monitored in environmental water. Memory accomodates up to 10 user-defined programs with up to 12 data points each.
Colorimeter features automatic wavelength selection, advanced reagent blank correction, and calibration curve adjustment. Analysis can be performed with user-set or pre-set reaction times through the built-in timer. Display reads results as concentration, absorbance, and percent transmittance.
Store and recall up to 99 measurements with date, time, test parameter, program number, concentration, absorbance, % transmittance, and sample number information. Data can be downloaded to a Microsoft® Windows® compatible spreadsheet or word processing program via HachLink™ software and data transfer adapter (both sold separately).
Colorimeter features a waterproof, dustproof, and chemically resistant housing.
What's Included:
two sample cells, 16-mm COD/Test' N Tube (TNT) tube adapter, four AA batteries, and instruction manual. Order printer and other accessories separately.
Product Type Color/Gloss Meters
Test parameters Multiparameter
Display LCD
Operating temperature 0 to 50°C (32 to 122°F)
Wavelengths Automatic
Waterproof Yes
Datalogging Yes
Dimensions 3.375" W x 9.375" H x 1.875" D
Model 48460-00
Brand Hach
Manufacturer number 48460-00
Power Four 1.5V AA batteries
Accessories
Qty/Price
Availability
at $269.50 USD/EACH
Usually ships in 14 days.
HachLink software
EW-99573-51
at $549.00 USD/EACH
Usually ships in 5 days.
at $120.50 USD/EACH
Usually ships in 6 days.
Hard-sided case
EW-99573-53
at $76.00 USD/EACH
In stock.
at $44.50 USD/PACKAGE of 6
In stock.
at $22.75 USD/EACH
Usually ships in 7 days.
at $21.00 USD/EACH
In stock.
at $21.00 USD/EACH
In stock.
at $10.75 USD/EACH
Usually ships in 6 days.
at $33.10 USD/EACH
Usually ships in 6 days.
Test tube rack
EW-99574-54
at $67.50 USD/EACH
Usually ships in 5 days.
at $860.00 USD/EACH
In stock.
at $860.00 USD/EACH
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title