Hach Total Phosphorus(USEPA) Test Kit

EW-99574-18
$72.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 7 days.
Qty

  • Environmental test kits for your Hach Colorimeter
Order the test kits for the Hach colorimeter to complete your water testing protocol. More test kits are available—if you don't see the one you need, call our Application Specialists for availability at 800-323-4340 x 6012.
All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120VAC 0r 99574-65, 240 VAC shown below).
Requires COD reactor listed below.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Water Testing Kit
Tests/kit 50
Range 0 to 3.5 ppm
Manufacturer number 27426-45
Brand HACH
Model 27426-45

Accessories
Qty/Price
Availability

at $13.00 USD/EACH

Usually ships in 6 days.

at $39.00 USD/EACH

Usually ships in 6 days.
Test tube rack
EW-99574-54

at $81.00 USD/EACH

Usually ships in 5 days.

at $1,025.00 USD/EACH

In stock.

at $1,025.00 USD/EACH

Usually ships in 7 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    I am looking for a TP test we can use in the field. Does this kit require the COD reactor?
    Asked on 1/19/2010 by Mary from Milwaukee, WI

    1 answer

    • Staff Reviewer

      A:

      Yes, this test requires a COD reactor like 99574-60.

      Answered on 10/29/2010 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title