log in or register
Hach digital titrator
EW-24908-00
$154.00 USD/EACH
  • Faster and more accurate than a conventional burette
  • Portable, precision dispensing with automatic calculation of titrant - works in field or lab settings
  • Interchangeable, sealed cartridges with concentrated titrants save time and eliminate glassware cleanup
The Hach digital titrator produces repeatable results through the use of concentrated titrants packaged in compact interchangeable cartridges. To titrate, simply attach a cartridge to the titrator and insert a delivery tube. Each turn of the delivery knob dispenses a calculated amount of titrant. The numerical counter records the volume. When the end point is reached, calculate the sample concentration using the number displayed on the counter. Any remaining reagent may be capped and stored for future use.
What's Included:
plastic case and five straight delivery tubes. Order interchangeable titrator cartridges separately.
Product Type Digital Titrators
Dispensing accuracy 800 digits/mL or 0.00125 mL/digit
Accuracy ±1%
CE Compliance Yes
Brand Hach
Manufacturer number 16900-01
Model 16900-01
Accessories
Qty/Price
Availability
at $12.10 USD/PACKAGE
Usually ships in 6 days.
at $8.50 USD/PACKAGE
Usually ships in 6 days.
at $20.00 USD/EACH
Usually ships in 5 days or sooner.
at $17.75 USD/EACH
Usually ships in 6 days.
at $18.50 USD/EACH
In stock.
at $20.50 USD/EACH
Usually ships in 10 days.
at $20.50 USD/EACH
Usually ships in 3 days.
at $20.50 USD/EACH
Usually ships in 6 days.
at $19.40 USD/EACH
Usually ships in 6 days.
Technical Resources
  • 24908-00.pdf
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title