Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register
Home  > 
Discontinued Items >
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.

Hach sensION 5 Waterproof Conductivity Meter

Hach  |   Item# EW-19604-00

$665.00 USD/EACH


  • Chemical-resistant, dustproof, and waterproof meter for your most demanding applications
  • Ultra-fast response—get your result within seconds
  • IP67-rated housing—waterproof, rugged, and it floats!
  • Large backlit LCD with anti-glare coating
Four-band conductivity cell (included) covers the entire measuring range of the meter—reduces the effects of polarization. Stores up to 99 data points; download data to a printer or computer via the docking station 59005-50 (order separately).
What's Included:
sensION™ four-band conductivity cell (19604-50), beakers, and batteries.
Product Type Conductivity Meter
Conductivity accuracy ±0.5% FS (first three ranges), ±1% FS (upper range)
Conductivity range 0 to 199.9 µS; 200 to 19,999 µS; 2 to 19.99 mS; 20 to 199.9 mS
Conductivity resolution 0.1 µS; 0.01 mS; 0.1 mS
Range (Conductivity) 0 to 199.9 uS, 200 to 19,999 uS; 2 to 19.99 mS, 20 to 199.9 mS
Range (TDS) 0 to 50,000 mg/L
Range (Salinity) 0 to 43 g/L
Range (Temperature °C) - 10 to 110
Salinity resolution 0.1 g/l
Resolution (Temperature) 0.1 C
Resolution (Conductivity) 0.1 µS; 1 µS, 0.01 mS, 0.1 mS
Resolution (TDS) 3 significant digits
Salinity accuracy ±0.1 ppt (within applicable temperature range)
Resolution (Salinity) 0.1 g/L
TDS range 0 to 50,000 mg/l
Accuracy (Temperature) +/- 0.5 C
TDS resolution 3 s.d.
Accuracy (Conductivity) +/- 0.5% full-scale
Accuracy (TDS) +/- 0.5% full-scale
Accuracy (Salinity) +/- 0.1 ppt
Temperature compensation Automatic
Temp range -110 to 110 °C
Display LCD
Interface RS-232
Waterproof Yes
Power Four AA batteries (included); 110 VAC with docking station 59005-50; 220 VAC with docking station 59005-55
Datalogging (points) 99
Brand Hach
Dimensions (" W) 3-7/16
Dimensions (" H) 8-11/32
Dimensions (" D) 1-21/32
Temp resolution ±0.1°C
Temp accuracy ±0.5 °C
Output One-way RS-232
Battery four AA batteries; 110 VAC with docking station 59005-50; 220 VAC with docking station 59005-55
TDS accuracy ±0.5% FS
Cell K = 0.45 cm-1


at $9.25 $7.40 USD/PACKAGE of 4

In stock.

at $474.00 USD/EACH

Usually ships in 8 days.

at $126.00 USD/EACH
Usually ships in 10 days.

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

2 Questions | 2 Answers
Displaying questions 1-2
  • Q:

    How often should the meter be calibrated?
    Asked on 5/31/2011 by Anonymous

    1 answer



      The frequency of calibration will depend on your accuracy needs and frequency of use. If accuracy is of major concern, I suggest calibrating once a day, before taking your measurements. If your accuracy requirements are not as strict, you could easily move to a once a week calibration schedule.

      Answered on 6/3/2011 by Josh from Technical Support
  • Q:

    How do i calibrate the salinity meter? Is there buffer solution i can purchase?
    Asked on 7/8/2010 by Anonymous

    1 answer

    • Staff Reviewer


      Yes, there are conductivity and salinity calibration standards. The most common conductivity/salinity standards are part numbers 00653-47 (215 ppm NaCl), 00653-18 (745 ppm NaCl). There are others that may work better for the range you are working in though. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).

      Answered on 7/9/2010 by Jen from Technical Support
Displaying questions 1-2

Do you have a question about this product? 

Product Reviews