United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
Hand held digital vibration meter kit with Metric units
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-98861-05
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-66
NIST-Traceable Calibration for Vibration
$155.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Vibration Measurement
>
Handheld Digital Vibration Meter Kits
Share
|
Hand-held digital vibration meter kit with Metric units
EW-98861-05
$1,848.00 USD/EACH
Usually ships in 2 days.
Qty
:
Add to Cart
Save to shopping list
Measures in four selectable ranges
Versatile unit provides high-accuracy measurements
These meters measure displacement, velocity, and acceleration in four suitable ranges. Easy measurements with the simple touch of a button. Measurements are displayed on large LCD.
What's Included:
meter, vibration pickup, pickup cable, magnetic pickup base, calibration document supplied by the manufacturer, four AA batteries, and carrying case.
Specifications
Vibration range
Acceleration:
0.001 to 1999 µm
Displacement:
0.001 to 1999 mm/sec
Velocity:
0.001 to 199 g's
Frequency range
26 to +300,000 rpm (0.4 to 5000 Hz)
Display
LCD
Low battery indicator
Yes
Dimensions
4"W x 7 3/4"H x 1 3/4"D
(10.2 x 19.7 x 4.4 cm)
Power
Four AA batteries (included)
Accuracy
±2.5% full scale
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration for Vibration
EW-17101-66
at $155.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Vibration Sensor Selection Guide
Trouble Shooting Industrial Accelerometer Installations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title