United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
Hand operated siphon pump 5 GPM Polyethylene
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06422-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Drum and Hand Pumps
>
Hand-Operated Drum Pumps
>
Hand-Operated Siphon Drum Pumps
Share
|
Hand-operated siphon pump, 5 GPM, Polyethylene
EW-06422-20
$14.50 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Economical siphon pumps are ideal for a variety of fluids
Adaptable to most pails and drums
Siphon pumps are inexpensive, disposable hand pumps. Model 06439-00 features a squeeze-bulb action while models 06422-20 and 06439-10 feature a bellows action for quick and easy emptying of containers. Model 07079-70 creates a siphon with a few strokes of the pump.
Specifications
Product Type
Hand-Operated Siphon Pumps
Tube OD (in.)
0.75
Wetted parts
Polyethylene
Max temperature (º C)
140
Max viscosity
500 SSU
Adapter descriptions
2" IPS Bung
Dimensions
44"L x 1" dia
Accessories
Qty
/
Price
Availability
Steel screwin adapter, 1-3/4" NPT(F)
EW-07079-10
at
$14.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Flex spout adapter, polyethylene
EW-07079-15
at
$11.00 USD/EACH
In stock.
Add to Cart
Bung converter kit converts a Mauser course bung thread or standard course buttress to 2" NPT
EW-07079-17
at
$19.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Closed System Technology Drives the Trend Toward Safer, More Cost-Efficient Chemical Dispensing
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title