United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
EW-09411-50
Handheld adhesive roll mop; 9" wide
Click to enlarge
More Product Details
Specifications
Replacement parts
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-09411-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Cleaning Supplies
>
Mops
>
Adhesive Roll Mops
Share
|
Handheld adhesive roll mop; 9" wide
EW-09411-50
$62.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Cut contamination with these adhesive roll mops. Mops have adhesive coated surfaces to remove debris and lint from glass, plastic, metal, and other problem surfaces. Choose from handheld (3"W or 9"W) or telescoping mops.
Handheld models 09411-40 and -50 are perfect for cleaning work counters, windows, or anywhere quick cleaning is needed in a confined space. Telescopic model 09411-60 has a 48"L handle that extends to 7 feetideal for hard to reach areas like walls, ceilings, floors, and corners.
Each roll mop has 20 disposable adhesive sheets; peel away the top layer to expose a fresh surface. Sheets are perforated every 14-3/8" for easy access to a fresh sheet. Refill rolls for each mop are sold separately.
Specifications
Description
Hand-held
Size
9"
Replacement parts
Qty
/
Price
Availability
Refill for roll mop 09411-50
EW-09411-53
at $30.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title