Hazardous Material Undercounter Refrigerator, 6 cu ft
YO-44765-70
$3,087.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 3 days.
Qty
  • Durable commercial cooling at an affordable price
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.Hazardous Location Models are for use where explosive conditions could potentially exist external to the cold storage unit. These units contain a specially designed compressor with all electrical components enclosed to prevent accidental ignition of refrigerator contents. These products have a UL listing for Class 1, Group C & D applications and must be hard-wired to voltage source.
What's Included:
Commercial power cord and plug, except for explosion-proof models which require hard-wiring.
Product Type Refrigerator
Capacity (cu ft) 6.1
Capacity (Liters) 173
Dimensions (interior) 28"H x 21"W x 20 3/4"D
Dimensions (overall) 34"H x 23 7/8"W x 26"D
Max Temp - Refrigerator (º F) 45
No. of doors 1
No. of shelves 2 interior, 3 door
Power (Hz) 60
Power (VAC) 115
Brand Marvel
Manufacturer number 6EAR0001
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  • 13-14 Spanish Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title