log in or register
Heat sealer, 12", 115 VAC
EW-03018-10
$136.25 USD/EACH
  • Create your own sealed bags
  • For added convenience some models include a cutter
Use new heat sealers to make an 8 or 12" seal on virtually any plastic sheet, bag, or flat tubing—seals PVC polyethylene, polypropylene, PLIOFILM®, SARAN®, and other transparent plastics. Simply set timing knob, press handle down on plastic, and remove when timing light indicates timing cycle is complete. Maximum temperature is 752°C (400°C).
Use the polyethylene bags (below) to make almost any size package quickly and easily. Customize your bags with the polyfilm polyethylene flat tubing. Simply roll off the amount of polyethylene flat tubing, seal at the correct length, and cut off your bag. Create your own workstation with heat sealers with cutters, optional roll holder 03015-40, and flat tubing (below). Heat sealers include a 6-ft , three-wire cord (115 VAC models include plug). Heat sealers for 8" seal measure 14½"L x 3-1/4"W x 7"H; heat sealers for 12" seal measure 18½"L x 3¼"W x 7"H
Product Type Standard heat sealer
Max temperature (º F) 750
Max temperature (º C) 399
Seal (" W) 0.08
Seal (" L) 12
Power (VAC) 115
Max temperature 750°F (399°C)
Dimensions 18.5 in L x 3.25 in W x 7 in H
(47.0 x 8.3 x 17.8 cm)
Accessories
Qty/Price
Availability
at $20.00 USD/EACH
In stock.
at $33.75 USD/EACH
Usually ships in 8 days.
at $265.00 USD/ROLL
In stock.
Bags Ldpe 3x5 1000/pk
EW-06505-02
at $43.00 USD/PACKAGE
In stock.
Bags Ldpe 3x8 1000/pk
EW-06505-04
at $54.00 USD/PACKAGE
Call 1-888-358-4705 for availability.
Bags Ldpe 4x5 1000/pk
EW-06505-06
at $47.00 USD/PACKAGE
In stock.
Bags Ldpe 5x7 1000/pk
EW-06505-08
at $63.00 USD/PACKAGE
In stock.
Bags Ldpe 6x9 500/pk
EW-06505-10
at $54.00 USD/PACKAGE
In stock.
Bags Ldpe 6x18 500/pk
EW-06505-12
at $98.00 USD/PACKAGE
In stock.
Bags Ldpe 8x10 500/pk
EW-06505-14
at $80.00 USD/PACKAGE
Call 1-888-358-4705 for availability.
at $100.00 USD/PACKAGE
In stock.
at $112.00 USD/PACKAGE
In stock.
at $212.00 USD/PACKAGE
In stock.
at $278.00 USD/PACKAGE
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title