dummy
EW-03018-15
Usually ships in 8 days.
Qty:
- Create your own sealed bags
- For added convenience some models include a cutter
Use new heat sealers to make an 8 or 12" seal on virtually any plastic sheet, bag, or flat tubing—seals PVC polyethylene, polypropylene, PLIOFILM®, SARAN®, and other transparent plastics. Simply set timing knob, press handle down on plastic, and remove when timing light indicates timing cycle is complete. Maximum temperature is 752°C (400°C).
Use the polyethylene bags (below) to make almost any size package quickly and easily. Customize your bags with the polyfilm polyethylene flat tubing. Simply roll off the amount of polyethylene flat tubing, seal at the correct length, and cut off your bag. Create your own workstation with heat sealers with cutters, optional roll holder 03015-40, and flat tubing (below). Heat sealers include a 6-ft , three-wire cord (115 VAC models include plug). Heat sealers for 8" seal measure 14½"L x 3-1/4"W x 7"H; heat sealers for 12" seal measure 18½"L x 3¼"W x 7"H
| Product Type |
Standard heat sealer |
| Max temperature (º F) |
750 |
| Max temperature (º C) |
399 |
| Seal (" W) |
0.2 |
| Seal (" L) |
12 |
| Power (VAC) |
230 |
| Max temperature |
750°F (399°C) |
| Dimensions |
18.5 in L x 3.25 in W x 7 in H (47.0 x 8.3 x 17.8 cm) |
at $20.00 USD/EACH
In stock.
at $33.75 USD/EACH
Usually ships in 8 days.
at $265.00 USD/ROLL
In stock.
at $43.00 USD/PACKAGE
In stock.
at $54.00 USD/PACKAGE
Call 1-888-358-4705 for availability.
at $47.00 USD/PACKAGE
In stock.
at $63.00 USD/PACKAGE
In stock.
at $54.00 USD/PACKAGE
In stock.
at $98.00 USD/PACKAGE
In stock.
at $80.00 USD/PACKAGE
Call 1-888-358-4705 for availability.
at $100.00 USD/PACKAGE
In stock.
at $112.00 USD/PACKAGE
In stock.
at $212.00 USD/PACKAGE
In stock.
at $278.00 USD/PACKAGE
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection