United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 15
of
19 >
Representative photo only
Heavy duty SS cart with guard rails 21 x 33 3 shelves 650 lb capacity
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-00946-15
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Handling and Storage
>
Hand & Platform Trucks
>
Stainless Steel Laboratory Carts
Share
|
Heavy-duty SS cart with guard rails; 21" x 33", 3 shelves, 650 lb capacity
EW-00946-15
$1,650.00 USD/EACH
Usually ships in 12 days.
Qty
:
Add to Cart
Save to shopping list
Built to accommodate oversized items
Enameled steel deck and angle subframe make these trucks very durable. Trucks 47350-00 and -10 have 5" diameter non-marking polyeurethane casters. High-capacity trucks 47350-20 and -30 have 6" molded-on rubber casters. Handle extends 28" above deck.Designed to reduce body strain. Vertical push handles keep hands in a comfortable position, reducing stress on arms and wrists. Padding on the handles (all models except 47520-00 and -02) absorbs vibrations and ensures a good grip. The bottom shelf is placed 20" from the floor to prevent potential back strain.
Specifications
Product Type
High Capacity Carts
Dimensions
22.375 in x 38.625 in x 37.25 in
Dimensions (" W)
38.63
Dimensions (" H)
37.25
Dimensions (" L)
22.38
No. of shelves
3
Shelf clearance
9 3/8"
Capacity (kg)
295
Caster size
5 in
Capacity (lbs)
650
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title