United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 25
of
31 >
EW-17710-56
Save over $1150 with this centrifuge bundle!
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-17710-56
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Centrifuges
>
General Purpose Centrifuges
>
Thermo Scientifc Heraeus Centrifuges
>
Thermo Scientific Heraeus Promotion
Share
|
Heraeus Megafuge 16 120V Clinical Bundle 28/32
EW-17710-56
$5,190.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Includes:
NCI SKu Heraeus Megafuge 16, 120v
17709-10 TX-200 rotor
17709-16 200 ml Round buckets (set /4) - for TX-200 rotor
17709-18 Sealing caps for TX-200 bucket (set /4, includes grease)
NCI Sku Set of 4 adapters 7 x 15/10 ml vac
NCI Sku Set of 4 adapters 8 x 5/7 ml
Specifications
Manufacturer number
75310885
Technical Resources
Related technical articles (Roll over for summary)
Tech Challenge: Centrifugation of Oil Samples in the Field
High Throughput Maxi-Prep Using the New Thermo Scientific General Purpose Centrifuge with Feberlite Rotors
Cole-Parmer Introduces Enhancements to Life Science Offering
Show All...
Isolating Genomic DNA from Whole Blood
Common Centrifuge Terms
Basics of Centrifugation
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title