log in or register
Hf Acid Eater Spill Kit; 5 Gal.; 1 Pack/case
EW-86295-04
$245.00 USD/EACH
Usually ships in 9 days.
Qty
  • Neutralizes Hydrofluoric Acid into safe Organic salts
Unlike caustic soda and soda ash, which forms poisonous bi-fluoride, HF Acid Eater completely destroys the hydrofluoric acid polymer. This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda. pH indicator additive eliminates the need for additional pH strips taking out the guess work. HF Acid Eater is nonhazardous and noncorrosive, and is excellent for a variety of uses and applications. Apply on horizontal, vertical, and perforated surfaces. Also neutralizes vapors and reduces odors.
Description HF ACID EATER SPILL KIT
Qty/cs 1 PACK/CASE
Quantity 2 GAL
Manufacturer number 2901-005
Model 2901-005
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title