United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
Representative photo only
Hinged Lid Sample Containers PP 1 oz 1300 pk
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-66098-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Containers
>
Plastic Containers
>
Hinged-Lid Sample Containers, PP
Share
|
Hinged-Lid Sample Containers, PP, 1 oz, 1300/pk
EW-66098-30
$433.00 USD/PACKAGE of 1300
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$389.70/PACKAGE of 1300
30
15%
$368.05/PACKAGE of 1300
120
20%
$346.40/PACKAGE of 1300
In stock.
Qty
:
Add to Cart
Save to shopping list
Hinged cap design allows you to easily close the lid with one finger, reducing chances of sample contamination
Made from polypropylene (PP); ideal for storing liquids or solids and for metals analysis. Containers are FDA compliant - for food. When containers are properly closed, your sample won't spill or leak in any position.
Autoclavable.
Specifications
Product Type
Sample Containers
Material
Polypropylene (PP)
Capacity (mL)
30
Capacity (oz)
1
Height (in.)
1-1/2
Height (cm)
3.8
Top OD (cm)
3.8
Top OD (in.)
1-1/2
Qty/pk
1300
Accessories
Qty
/
Price
Availability
1 1/4" Label for 1.5" Diameter Container. White, 1000/Roll
EW-06098-01
at
$42.00 USD/PACKAGE of 1000
Usually ships in 6 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Physical Properties of Plastics
Physical Properties of Glass
Physical Properties of Stainless Steel
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title