log in or register
Honeywell Impact Multigas Monitor
EW-81948-40
$582.00 USD/EACH
Usually ships in 10 days.
Qty
  • Replaceable sensor cartridges minimize maintenance downtime and cost
  • Increasingly rapid flashing, ultra-bright alarm LEDs
  • Three levels of instantaneous alarm
  • Time-weighted average alarms on toxic
The Impact multigas monitor is capable of monitoring up to four gases. Select 12-month disposable cartridges based on gases present. Check gas prior to entering a confined space using a drop line, hand aspirator, or optional pump. Patented Surecell sensors ensure stable, accurate, and reliable operation. A large, multi-lingual, backlit display with intuitive menu system simplifies use and minimizes operator training. The durable, rugged impact-resistant housing is waterproof to IP67 rating, yet small and lightweight for use in tight spaces. Meters feature a periodic “confidence” flash and beep, and powerful two-tone audible alarm.
Approval: UL and CSA Class I, Div I, Groups A, B, C, and D
Note: Cartridges are required and sold separately.
What's Included:
belt clip, flow adapter, dry cell battery holder, and batteries.
Product Type Multigas Monitors/Detectors
Power two dry cell batteries (included)
Battery life 17 hours
Instrument wt 18 oz (0.52kg)
Dimensions 3.375 in W x 2 in H x 5.375 in D
Brand Honeywell
Manufacturer number 2302B20008UE
Accessories
Qty/Price
Availability
at $185.00 USD/EACH
Usually ships in 14 days.
at $225.00 USD/EACH
Usually ships in 8 days.
at $225.00 USD/EACH
Call 1-888-358-4705 for availability.
at $280.00 USD/EACH
Usually ships in 8 days.
at $200.00 USD/EACH
Call 1-888-358-4705 for availability.
at $216.00 USD/EACH
Call 1-888-358-4705 for availability.
at $100.00 USD/EACH
Usually ships in 7 days.
at $313.00 USD/EACH
Call 1-888-358-4705 for availability.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title