dummy
EW-81948-41
Usually ships in 15 days.
Qty:
- Replaceable sensor cartridges minimize maintenance downtime and cost
- Increasingly rapid flashing, ultra-bright alarm LEDs
- Three levels of instantaneous alarm
- Time-weighted average alarms on toxic
These multigas monitors are capable of monitoring up to four gases (multigas cartridges are sold separately). Check gas prior to entering a confined space using a drop line, hand aspirator, or built-in sampling pump. Patented Surecell sensors ensure stable, accurate, and reliable operation. A large, multi-lingual, backlit display with intuitive menu system simplifies use and minimizes operator training. The durable, rugged impact-resistant housing is waterproof to IP67 rating, yet small and lightweight for use in tight spaces. Meters feature a periodic "confidence" flash and beep, and powerful two-tone audible alarm.
Impact Pro features a built-in sampling pump, vibrating alarm, and multilingual menu. Link two Impact Pro detectors together using the optional Safelink cable to safely monitor personnel in a space.
Note: Sensor cartridges are required and sold separately.
What's Included:
belt clip, flow adapter, dry cell battery holder, and batteries. Order cartridges separately.
| Product Type |
Multigas Monitors/Detectors |
| Power |
two dry cell batteries (included) |
| Battery life |
17 hours |
| Instrument wt |
18 oz (0.52kg) |
| Dimensions |
3.375 in W x 2 in H x 5.375 in D |
| Brand |
Honeywell |
| Manufacturer number |
2302B20009UE |
at $185.00 USD/EACH
Usually ships in 14 days.
at $225.00 USD/EACH
Usually ships in 8 days.
at $225.00 USD/EACH
Call 1-888-358-4705 for availability.
at $280.00 USD/EACH
Usually ships in 8 days.
at $200.00 USD/EACH
Call 1-888-358-4705 for availability.
at $216.00 USD/EACH
Call 1-888-358-4705 for availability.
at $100.00 USD/EACH
Usually ships in 7 days.
at $313.00 USD/EACH
Call 1-888-358-4705 for availability.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection