United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-89550-00
Honeywell
®
UDC 700 Universal-Input Controller, 90-264V
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-89550-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-18
NIST-Traceable Calibration; Panel Meters
$75.00 USD/EACH/EACH
Usually ships in 10 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Panel Meters
>
Honeywell UDC 700-Series Temperature/Process Controller
Share
|
Honeywell UDC 700 Universal-Input Controller, 90-264V
EW-89550-00
$240.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Compact 1/32 DIN size gives you high performance in a small package
Universal-Input
This controller features automatic, hands-off PID tuning or manual tuning. The NEMA 4/IP66 front face protects against dust and water. All input types are field configurable.
For additional features, see "Customize Your UDC 700 Controller" at right.
Specifications
Product Type
Universal controllers
Power (VAC)
90 to 264
DIN size
1/32"
Output (1)
Relay: SPST, isolated, rated @ 2A @ 120 V or 240 V; resistice load
Display
4-digit red LED, 0.4"H
Relay output
SPST
Operating conditions
32 to 131°F (0 to 55°C)
Panel cutout
1.79"W x 0.81"H
Alarm
nonisolated SSR driver rated @ >10VDC, 500
minimum (50 mA max)
Enclosure
NEMA 4 (IP66)
Dimensions
2"W x 1"H x 4-5/16"D
Brand
Honeywell
Manufacturer number
DC701-0-0-0-0
Model
DC701-0-0-0-0
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Panel Meters
EW-17090-18
at $75.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Customize Your UDC 700 Controller
Related technical articles (Roll over for summary)
Process/Panel Meter Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title