dummy
EW-19600-20
- Large LCD shows temperature and pH, mV, or conductivity simultaneously
- Self diagnostic function ensures reliable measurements
- Nonvolatile memory stores all calibration data and up to 300 measurements
Waterproof meters feature IP67-rated housing for field or lab use. Download data using RS-232 capabilityorder cable 99591-70 separately. Order digital conductivity simulator 19600-30 to calibrate meter without having to use liquid standards. D-54 meter measures pH, temperature, ORP, and conductivity.
What's Included:
soft-side case with strap, and batteries. Order electrodes separately.
| Product Type |
Multiparameter Meters |
| Dimensions (" W) |
3 |
| Dimensions (" H) |
7 |
| Dimensions (" D) |
2-1/4 |
| Range (pH) |
0.00 to 14.00 |
| Range (mV) |
±1999 |
| Range (Conductivity) |
K = 0.1, 0.0µS to 19.99 mS K = 1.0, 0.00 mS to 199.9 mS K = 10, 0.00 mS to 1999 mS |
| Range (Temperature °C) |
0 to 100 |
| Resolution (pH) |
0.01 |
| Resolution (Temperature) |
0.1°C |
| Resolution (mV) |
1 |
| Resolution (Conductivity) |
0.05% full scale |
| Accuracy (pH) |
±0.01, ±1 digit |
| Accuracy (Temperature) |
±0.1°C ±1 digit |
| Accuracy (mV) |
±1 mV ±1 digit |
| Accuracy (Conductivity) |
±0.5% full scale, ±1 digit |
| Temperature compensation |
Automatic |
| Display |
LCD |
| Interface |
RS-232 |
| Waterproof |
Yes |
| Buffer recognition |
NIST, DIN |
| Power |
One 9 V battery (included) |
| Datalogging (points) |
300 |
| Brand |
Horiba |
| Model |
352734-1 |
at $710.00 USD/EACH
Call 1-888-358-4705 for availability.
at $460.00 USD/EACH
Call 1-888-358-4705 for availability.
at $430.00 USD/EACH
In stock.
at $470.00 USD/EACH
Usually ships in 9 days.
at $510.00 USD/EACH
Call 1-888-358-4705 for availability.
at $220.00 USD/EACH
Usually ships in 10 days or sooner.
at $160.00 USD/EACH
In stock.
at $37.00 USD/EACH
Call 1-888-358-4705 for availability.
at $250.00 USD/EACH
Usually ships in 6 days.
at $115.00 USD/EACH
Usually ships in 9 days.
at $115.00 USD/EACH
Usually ships in 9 days.
at $366.00 USD/EACH
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334