Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Horiba Oil Content Analyzer, 115 V

EW-82900-00
$7,000.00 USD/EACH

Usually ships in 7 days.
Qty

  • Routine analysis of oil residuals
The Horiba OCMA-350 analyzer simplifies measurement of oil in water or soil for a wide variety of applications. Use this precision instrument for surveying water quality and hazardous waste sites, industrial wastewater monitoring, post-cleaning discharge analysis, and measuring adherent oil residuals on textiles and metal parts.

Use S-316 solvent to quantitatively extract sample. The oil / solvent solution is then simply transferred to the OCMA-350 analyzer for single-button measurement. Analysis is based on infrared (IR) spectral data, with peak integration (at approximately 3.4 µm) to determine oil concentration. S-316 solvent is non-absorbant at this IR wavelength, so only oil is measured. Accurately measures oil down to 0.1 ppm. Auto-calibrates after initial standard reading.

Use built-in time stamping to document results. Read results on LCD or transfer data using parallel printer port or to computer via RS-232. S-316 solvent can be recycled using Solvent Reclaimer SR-300 (82900-20), sold separately below.

What’s included: Quartz cell with cap, power cord, B-heavy oil (standard), 2 amp fuse, 10 mL syringe and 25 µL syringe.
Product Type Petroleum Testing Spectroscopes
Photometric readout 0 to 200 mg/L, ranges 0 to 9.9, 10 to 99.9 or 100 to 200
0 to 1000 mg/kg, ranges 0 to 9.99, 10 to 99.9 or 100 to 1000
0 to 1 Absorbance, range 0 to 1000
Sensitivity 1% of scale lower limit
Detector Pyroelectric
Output RS-232C and centronics printer port
Pathlength 200 mm quartz cell
Display LCD
Dimensions (cm) 25cm x 20cm x 28.5cm
Model 350751
Brand Horiba

Accessories
Qty/Price
Availability

at $420.00 USD/EACH

In stock.

at $1,855.00 USD/EACH

Usually ships in 12 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title