United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
Horiba Oil Content Analyzer 230 V
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-82900-05
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Specialty spectrophotometers
>
Horiba Oil Content Analyzer
Share
|
Horiba Oil Content Analyzer, 230 V
EW-82900-05
$7,000.00 USD/EACH
Usually ships in 12 days.
Qty
:
Add to Cart
Save to shopping list
Routine analysis of oil residuals
The Horiba OCMA-350 analyzer simplifies measurement of oil in water or soil for a wide variety of applications. Use this precision instrument for surveying water quality and hazardous waste sites, industrial wastewater monitoring, post-cleaning discharge analysis, and measuring adherent oil residuals on textiles and metal parts.
Use S-316 solvent to quantitatively extract sample. The oil / solvent solution is then simply transferred to the OCMA-350 analyzer for single-button measurement. Analysis is based on infrared (IR) spectral data, with peak integration (at approximately 3.4 µm) to determine oil concentration. S-316 solvent is non-absorbant at this IR wavelength, so only oil is measured. Accurately measures oil down to 0.1 ppm. Auto-calibrates after initial standard reading.
Use built-in time stamping to document results. Read results on LCD or transfer data using parallel printer port or to computer via RS-232. S-316 solvent can be recycled using Solvent Reclaimer SR-300 (82900-20), sold separately below.
What’s included:
Quartz cell with cap, power cord, B-heavy oil (standard), 2 amp fuse, 10 mL syringe and 25 µL syringe.
Specifications
Product Type
Petroleum Testing Spectroscopes
Photometric readout
0 to 200 mg/L
, ranges 0 to 9.9, 10 to 99.9 or 100 to 200
0 to 1000 mg/kg
, ranges 0 to 9.99, 10 to 99.9 or 100 to 1000
0 to 1 Absorbance
, range 0 to 1000
Sensitivity
1% of scale lower limit
Detector
Pyroelectric
Output
RS-232C and centronics printer port
Pathlength
200 mm quartz cell
Display
LCD
Dimensions (cm)
25cm x 20cm x 28.5cm
Model
OCMA-350-220V
Brand
Horiba
Accessories
Qty
/
Price
Availability
Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer
EW-82900-10
at
$420.00 USD/EACH
In stock.
Add to Cart
Solvent reclaimer SR-300, for recycling Horiba Oil Content Analyzer extractant
EW-82900-20
at
$1,855.00 USD/EACH
Usually ships in 12 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title