Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Horiba PA-1000 Environmental Radiation Monitor

EW-50538-02
$1,731.00 USD/EACH

Usually ships in 5 days.
Qty

  • Measures environmental radiation (gamma rays) from 0.001 µSv/h
  • IP54 splash-resistant construction is ideal for use in wet locations
  • Lightweight compact design is perfect for use out in the field
This environmental radiation monitor allows you to easily measure minute levels of gamma rays.
Monitor natural environmental radiation emitted from the ground to the air, as well as from manmade objects such as concrete and buildings.

Available for sale only in the United States. A personal radiation detector is one of the many devices lab workers use to keep them safe. Exposure to radiation through contamination or irradiation can be a significant hazard. Even small amounts over a long time can increase the risk of cancer. In emergency situations, radiation can cause illness, burns, or even death. In nuclear disasters, both the radiation itself and the radioactive ash can be lethal. If a radiation dose is high and penetrates the body, Acute Radiation Syndrome (ARS) may occur. That’s why a personal radiation detector is used to prevent harm. A personal radiation detector monitors the environment to alert users to the presence of radioactive material. Depending on the type (gamma radiation detector, alpha radiation detector, radiation dosimeters), they can detect and measure types of radiation, levels of radiation, counts, accumulated dose, and dose rates. Cole-Parmer offers several low-level sensitive handheld and table-top units with varying features, including documentation capabilities, timers, and ergonomic design. General-purpose detectors are available with uncompensated halogen-quenched sensors for measuring alpha, beta, gamma, and X-ray radiation; select meters have energy-compensated sensors for increased sensitivity to gamma and X-ray radiation. Personal, pen-style dosimeters are also an option. If you have questions about purchasing a personal radiation detector contact a Cole-Parmer Technical Expert today.
What's Included:
Two AA batteries.
Product Type Environmental Radiation Monitor
Radiation detected Gamma rays
Sensitivity Min. 1000 counts per minute (1000 cpm) for 1 µSv/h
Range 0.001 to 9.999 µSv/h
Display 4-digit digital display (count value converted into µSv/h)
Energy range > than 150 keV
Sampling rate 60 seconds
Detector Scintillation
Accuracy ±10%
Brand Horiba
Model PA-1000
Power Two AA batteries

Accessories
Qty/Price
Availability

at $7.50 USD/PACKAGE of 4

In stock.




 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title