United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-68517-01
Humidity/Temperature USB Datalogger
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68517-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-24
NIST-Traceable Calibration; %RH/Temperature Datalogger
$175.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Recorders & Data Acquisition
>
Dataloggers
>
Temperature & Humidity Dataloggers
>
Extech Temperature and Humidity Datalogger
Share
|
Humidity/Temperature USB Datalogger
EW-68517-01
$220.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Datalogs up to 16,000 readings with USB interface for downloading to a PC
Triple LCD displays humidity, temperature and date (year, month and day)
Programmable from keypad or PC
Selectable data sampling rate
User programmable audible and visual alarms
Wall, desktop or tripod mount
Calibration via optional salt bottles
What's Included:
Windows
®
compatible software, USB cable, 110V AC adaptor and four AA batteries
Specifications
Product Type
Temperature/Humidity Dataloggers
CE Compliance
Yes
Brand
Extech
Manufacturer number
42280
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; %RH/Temperature Datalogger
EW-17030-24
at $175.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual
Related technical articles (Roll over for summary)
Tech Challenge: Temperature Monitoring of Food Products
Benefits of Universal Serial Bus (USB) for Data Acquisition
Benefits of Simultaneous Data Acquisition Modules
Show All...
Gotchas for USB Measurement
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title