dummy
EW-86509-00
Usually ships in 9 days.
Qty:
- Centrifugal motion draws sample at 40 L/min
- Ideal for air quality, clean area, and food and beverage applications
- Lightweight, compact design for portability and ease of use
The Hycon RCS air sampler deliversreliable reproducible results for applications assessing the quality of ambient air. Sampler operates on impaction principle, drawing airborne microorganisms onto an agar media strip for later incubation and analysis. Use keypad to adjust sample times: 0.5, 1, 2, 4, and 8 minutes; LED indicators provide visual indication of each sampling run. Rugged housing is chrome-plated brass with autoclavable, aluminum impeller blade and drum. Order accessory kit, batteries, power adapter, and agar strips separately.
| Power |
Four D batteries or optional adapter |
| Dimensions |
2"W x 14"H x 2"D |
| Manufacturer number |
1441640001 |
| Model |
940015 |
at $16.15 USD/PACKAGE of 4
In stock.
at $125.00 USD/BOX
Usually ships in 7 days.
at $84.00 USD/BOX
Call 1-888-358-4705 for availability.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.