Fisher Chemical A481212 Hydrochloric Acid (NF/FCC) (2.5 liter) CAS 7647-01-0

Representative image only.
Fisher Chemical Mfr # A481212 – Item # EW-88011-70
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — 36.5-38.0%
  • Color—To pass test
  • Residue on Ignition—≤0.008%
  • Bromide or Iodide—To pass test
  • Free Bromine or Chlorine—To pass test
  • Sulfate—≤0.5%
  • Sulfite—To pass test
  • Heavy Metals (as Pb)—≤1ppm
  • Iron—≤5ppm
  • Nonvolatile Residue—≤0.5%
  • Reducing Substances—≤0.007%
  • Oxidizing Substances (as Cl2)—≤0.003%
  • Total Organic Compounds (non-fluorine containing)—≤5mg/kg
  • Including Benzene—≤0.05mg/kg
  • Fluorinated Organic Compounds (total)—≤25mg/kg

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeFCC or NF
  • Empirical FormulaHCl
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • SynonymsMuriatic acid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight36.45
  • PackagingGlass
  • DOT InformationCorrosive; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive, Toxic
  • CAS Number7647-01-0 and 7732-18-5
  • UN NumberUN1789
  • DescriptionHydrochloric Acid, NF/FCC Grade; (2.5 liter)
CorrosiveStorage Code WhiteToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 36.5-38.0%
  • Color—To pass test
  • Residue on Ignition—≤0.008%
  • Bromide or Iodide—To pass test
  • Free Bromine or Chlorine—To pass test
  • Sulfate—≤0.5%
  • Sulfite—To pass test
  • Heavy Metals (as Pb)—≤1ppm
  • Iron—≤5ppm
  • Nonvolatile Residue—≤0.5%
  • Reducing Substances—≤0.007%
  • Oxidizing Substances (as Cl2)—≤0.003%
  • Total Organic Compounds (non-fluorine containing)—≤5mg/kg
  • Including Benzene—≤0.05mg/kg
  • Fluorinated Organic Compounds (total)—≤25mg/kg

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat