United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
24 >
EW-28710-00
IKA
®
Basic Rotary Evaporator With Vertical Uncoated Glassware, 115 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-28710-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Evaporators
>
Rotary Evaporator Systems
>
IKA Rotary Evaporators
Share
|
IKA Basic Rotary Evaporator With Vertical Uncoated Glassware, 115 VAC
EW-28710-00
$3,189.00 USD/EACH
Usually ships in 10 days.
Qty
:
Add to Cart
Save to shopping list
Safe, innovative design includes unique reversing feature for better evaporation
Five-year warranty
(excludes glassware and wearing parts)
Standard motorized lift automatically lifts flask out of bath in case of power failure
Smooth start reduces chance of splashing
Unique oblong bath design heats up faster than other leading brands
These rotary evaporators were designed and engineered for ease of use, while maintaining the highest level of safety. Motorized lift is standard, making for easier removal of the evaporating flask from the hot water/oil bath. The motorized lift automatically removes evaporating flask from bath in case of a power failure to minimize loss of your experiment. Adjustable lift setting to accommodate larger flasks.
The brushless DC-drive motor features reversing rotational function that is commonly used for drying powders. Motor features a smooth start at 100 rpm to minimize the chance of splashing. Digital rotation speed display is standard on all models, making for easier repetition of parameters. The unique bath design provides faster heating, allowing for quicker experiments. Nonexposed heating coils and integrated handles provide safe operation. Bath heats from 20 to 180°C, adding versatility by allowing use of water or oil.
Basic Evaporators
are ideal for routine distillations, and feature digital rotational speed setting and display from 20 to 270 rpm. Bath temperature is controlled by analog controller from 20 to 180°C.
What's Included:
rotary evaporator, water/oil heating bath, condenser set, evaporatoring and condensing flask (each 1 L), and 6-ft power cord.
Specifications
Product Type
Rotary Evaporator
Temp resolution
Analog
Bath temperature
Ambient to 180°C
Power (Hz)
50/60
Dimensions
19.7"W x 16.9"H x 16.1"D (50.0 x 43.0 x 41.0 cm)
Rotation speed range
20 to 270 rpm
Speed resolution
Digital display
Joint size
24/40 (evaporating)
Lift mechanism
Motorized
Condenser type
Vertical
Glass type
Uncoated
Brand
IKA
Power (VAC)
115
Manufacturer number
8023901
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title