United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
IKA Continuous feed grinding drive 115 VAC 60 Hz
Click to enlarge
More Product Details
Specifications
Reconditioned & Like New
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-04301-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
Continuous-Feed Grinding Mills
>
IKA Continuous-Feed Grinding Drive
Share
|
IKA Continuous feed grinding drive, 115 VAC/60 Hz
EW-04301-30
$2,824.00 USD/EACH
Usually ships in 10 days or sooner.
Qty
:
Add to Cart
Save to shopping list
Continuous feed allows unlimited sample sizes
Two grinding heads available for one driveinterchange heads for different applications
Adjustable speed control from 3000 to 6,500 rpm
Continuous dutygreat for inline applications
Surfaces that contact your samples are 304 SSideal for sanitary applications
Easy-to-clean stainless steel tray provides an ideal working surface
1-1/3 hp (990-watt) motor is completely sealed from the grinding chamber by a dust-tight seal
Constructed of heavy-duty cast aluminum, enameled finish, with a stainless steel tray. Order grinding heads separately.
Specifications
Product Type
Cutting and Grinding Materials/Media
Capacity
unlimited; continuous-feed
Speed
3000 to 6,500 rpm
Motor
1.333 hp
Power (Hz)
60
Dimensions
11.8125 in W x 17.625 in H x 12.625 in D
Power
115 VAC, 50/60 Hz
Brand
IKA
Manufacturer number
2836001
Power (VAC)
115
Reconditioned & Like New
Qty
/
Price
Availability
LIKE NEW 04301-30
EW
-
00319RW
at
$2,181.95 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title