United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-04300-30
Cutting-grinding head shown with drive. Components sold separately.
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-04300-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
Continuous-Feed Grinding Mills
>
IKA Continuous-Feed Grinding Head
Share
|
IKA Cutting-grinding head
EW-04300-30
$1,964.00 USD/EACH
Usually ships in 11 days.
Qty
:
Add to Cart
Save to shopping list
Continuous dutygreat for inline applications
Surfaces that contact your samples are 304 SSideal for sanitary applications
For use with IKA's Continuous-Feed Grinding Drive.
Cutting-grinding head is ideal for paper, vegetation, plastics, and materials with low-volume weight.
Specifications
Product Type
Cutting and Grinding Materials/Media
Speed
73.8 ft/s max (blade)
Feed Particle Size
15 mm max
Final Particle Size
250 µm
Brand
IKA
Manufacturer number
2870900
Accessories
Qty
/
Price
Availability
Sieve, 0.25 mm
EW-04300-33
at
$323.00 USD/EACH
In stock.
Add to Cart
Sieve, 0.5 mm
EW-04300-34
at
$323.00 USD/EACH
In stock.
Add to Cart
Sieve, 1.0 mm
EW-04300-35
at
$323.00 USD/EACH
In stock.
Add to Cart
Sieve, 2.0 mm
EW-04300-37
at
$323.00 USD/EACH
In stock.
Add to Cart
Sieve, 3.0 mm
EW-04300-38
at
$323.00 USD/EACH
Usually ships in 7 days or sooner.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title