United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-50705-00
RW 20 Mixer includes propeller, stand, and clamp
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-50705-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-03
Innocal NIST Calibration for Mixer
$115.00 USD/EACH/EACH
Usually ships in 13 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Laboratory Mixers
>
IKA Mixers
>
IKA RW 20 Digital Dual-Range Mixers
Share
|
IKA RW 20 Digital Dual-Range Mixer, 60 to 500/240 to 2000, 115 VAC
EW-50705-00
$875.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Great valuecomplete mixer package includes three free accessories: stand, clamp, and four-blade propeller!
Digital display makes reproducibility easy
Through-shaft design provides virtually limitless adjustment for impeller depth
Two speed ranges to suit a variety of mixing applications
Maintenance-free motor rated for continuous duty
Use this versatile, affordable mixer for liquid volumes up to five gallons and viscosities up to 10,000 cps (approximately the viscosity of honey). Select speed from 60 to 2000 rpm in two ranges for a variety of applications. Through-shaft design permits adjustment of shaft length and impeller depth—allows you to place or remove mixing vessels without repositioning the mixer.
The motor and drive unit are conveniently integrated into a compact aluminum and thermoplastic housing with easy-to-use controls. The quiet, fan-cooled motor features thermal overload shut-off and an efficient heat dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Mixer features stepless speed control. The 1⁄2-inch LCD shows rotational speed in rpm and enables you to precisely control and repeat process parameters.
What's Included:
chuck (accepts up to 3⁄8" [10-mm] dia shafts), 6-1⁄4"L x 1⁄2" dia mounting rod, four-blade propeller, mixer stand, clamp, and a 6-1⁄2-ft, three-wire power cord (115 VAC model includes plug).
Specifications
Product Type
Mixer
Chuck
10 mm
Input power
70 W
Min speed (rpm)
60
Max speed (rpm)
2000
Motor (hp)
0.04
Motor type
Brushless
Remote control
No
Max torque (in-oz)
259
Viscosity (cp)
10,000
Power (VAC)
115
Power (Hz)
60
Through-shaft
Yes
CE Compliance
Yes
Brand
IKA
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Mixer
EW-17050-03
at $115.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
More info
IKA
RW 20 Digital Dual-Range Mixer Brochure
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title