Ika 10 place multiposition stirring hot plate, 115V
YO-04671-61
$1,902.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 10 days or sooner.
Qty
  • Simulatenous stirring for consistency at each position
  • Models available up to 15 position for optimal use of lab space
These multiposition stirrers and stirring hot plates feature a powerful rotary drive providing consistent sample conditions across each position for volumes up to 400 mL. They are available in 5, 10, or 15 position configurations.
Stirrers have an easy-to-clean stainless steel surface which provides protection against liquid infiltration. Stirrers also include a removable polyurethane cover to protect the stirrer from scratches.
Stirring hot plates include an integrated temperature control plate. Precise temperature distribution on the heating plate allows for relieable series experminets. Use these units to heat only, stir only, or heat ands stir simultaneously.
Product Type Stirrer (Hot Plates)
Speed (rpm) (low) 0
Speed (rpm) (high) 1100
Max temperature (º C) 120
Top plate material Silicone
Top plate dimensions (in.) 7 in x 17.75 in
Dimensions 8 in W x 21.75 in D x 2.5 in H
Max stirring volume (Liters) 0.4
No. of stirring positions 10
Plug type U.S. std
Model RT 10 power IKAMAG
CE Compliance Yes
Brand IKA
Manufacturer number 2930501
Power 115V, 50/60 Hz
Calibrations
Qty/Price
Availability
at $160.00 USD/EACH
Usually ships in 8 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title