United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 9
of
12 >
EW-36136-20
Immersion Heater, Three PTFE Coated Rods, 6"l Heated Area, 500 Watts, 120 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36136-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Immersion Heaters
>
Multi-Purpose Immersion Heaters
>
Heaters for Hostile Environments
>
Multi-Element Imersion Heaters
Share
|
Immersion Heater, Three PTFE Coated Rods, 6"l Heated Area, 500 Watts, 120 VAC
EW-36136-20
$650.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Multi-Element Heaters Encased in 304 Stainless Steel or PTFE include a grounded solid-state heating element. Heaters are self-limiting for safer operation. PTFE-coated heaters are ideal for use in corrosive environments. The numbers of heating rods coming down from the polypropylene terminal head are dependent on the wattage of the heater: 300 Watt = 2 rods, 500 Watt = 3 rods, and 1000 Watt = 5 rods. Heaters include 3 ft wire and flexible conduit.
Specifications
Product Type
Multiple rods
Heated area
6 in L
Material
PTFE coated
Immersion depth
11 in
Temp range
Ambient to 450°F (232°C)
Watts
500
Power (VAC)
120
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Immersion Heater Selection Sheet
Related technical articles (Roll over for summary)
Cole-Parmer: Immersion Heater Selection Sheet
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title