United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 16
of
16
Representative photo only
Instant Blue Stain 1 liter
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-28570-59
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Electrophoresis Products
>
Reagents
>
ClearPAGE Electrophoresis Precast Gels
Share
|
Instant Blue Stain, 1 liter
EW-28570-59
$209.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Available for sale only in the United States
Quick and easy-to-use precast gels eliminate the need for traditional handcasting
Compatible with combination vertical systems (see "Related Product" ad at bottom)
Gel technology features a unique polymer matrix for exceptional protein seperation
Simple and reuseable saving time and money
Long shelf life saves your lab money
No need to throw away expired gels! These convenient precast gels have over a 12-month shelf life. No change in separation when stored properly during that period. Incredibly strong gels provide high resolution results and are highly resistant to tearing and cracking, even during the drying process. One-of-a-kind polyacrylamide gel with agarose formulation also reduces shrinkage and swelling, resulting in better resolution and more efficient transfer. Use precast gels with buffers sold under "Accessories"—they are not compatible with standard lab buffers. Large sample volumes will still product sharp bands. Cassette design allows longer pathlength, increasing resolution. All gels are 1-mm thick and use a red loading area for easy visualization of wells. Available in fixed or gradient acrylamide concentrations.
Specifications
Product Type
Electrophoresis Accessories
Manufacturer number
HG73010
Technical Resources
MSDS_28570-59.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
ChemicalsSpecial Ordering Information
Isolating Genomic DNA from Whole Blood
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title