United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 17
of
30 >
EW-86435-16
Kit 86435-06 is shown
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86435-16
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Arc Flash Products
>
Arc Flash Gloves
>
Salisbury by Honeywell Electrical Insulating Rubber Glove Kits
Share
|
Insulated Glove Kit, Class 0, Black, 11" length, size 9, with leather protector glove and glove bag
EW-86435-16
$131.00 USD/EACH
Usually ships in 14 days or sooner.
Qty
:
Add to Cart
Save to shopping list
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note:
In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.
What's Included:
insulating rubber glove, leather protector glove, and storage bag.
Specifications
Product Type
Gloves
Material
Rubber
Size
Medium
Length
11 in
Color
Black
Compliance
Class 0
Make
Salisbury
Manufacturer number
GK011B9
Model
GK011B9
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
If the Glove Fits: The Right Safety Glove for your Application
Safety Gloves Sizing Guide
Safety Gloves Selection Guide
Show All...
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Chemical Compatibility Chart for Safety Apparel
ABCs of DMMs
Safety Apparel Material Selection Guide
Cleanroom Garments
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title