Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Insulated Rescue Hook, 6 foot length

EW-86437-11
$385.00 USD/EACH

  • Heat-treated body hook has an 18" (45.7 cm) opening
  • Foam-filled, fiberglass-reinforced handle for superior electrical insulation
The rescue hook is an invaluable tool for any workplace. Withdraw an injured worker from a hazardous area, confined space, or near electrical cabinets and switchgear.
Material Foam-filled fiberglass-reinforced handle
Model 24401





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title